Assay Details
| Gene Symbol: LOC110345141 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021241428.1 | XP_021097087.1 | 3 - 4 | 717 | 108 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC110345141| Gene Symbol: LOC110345141 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021241428.1 | XP_021097087.1 | 3 - 4 | 717 | 108 | |
Set Membership: |
|