Assay Details
| Gene Symbol: Znf34 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004837660.2 | XP_004837717.1 | 3 - 4 | 751 | 58 | |
| XM_021249991.1 | XP_021105650.1 | 3 - 4 | 693 | 58 | ||
More Information
Set Membership: |
|

Znf34
Genomic Map