Assay Details
| Gene Symbol: Arhgef33 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004839329.2 | XP_004839386.1 | 15 - 16 | 1839 | 70 | |
| XM_004839332.2 | XP_004839389.1 | 15 - 16 | 1840 | 70 | ||
| XM_004839333.2 | XP_004839390.1 | 15 - 16 | 1839 | 70 | ||
| XM_021253745.1 | XP_021109404.1 | 16 - 17 | 1984 | 70 | ||
| XM_021253746.1 | XP_021109405.1 | 16 - 17 | 1982 | 70 | ||
| XM_021253748.1 | XP_021109407.1 | 16 - 17 | 1985 | 70 | ||
| XM_021253749.1 | XP_021109408.1 | 16 - 17 | 1984 | 70 | ||
| XM_021253750.1 | XP_021109409.1 | 15 - 16 | 1923 | 70 | ||
| XM_021253751.1 | XP_021109410.1 | 16 - 17 | 1985 | 70 | ||
| XM_021253752.1 | XP_021109411.1 | 16 - 17 | 1985 | 70 | ||
| XM_021253753.1 | XP_021109412.1 | 16 - 17 | 1985 | 70 | ||
| XM_021253754.1 | XP_021109413.1 | 16 - 17 | 1983 | 70 | ||
| XM_021253755.1 | XP_021109414.1 | 16 - 17 | 1985 | 70 | ||
| XM_021253756.1 | XP_021109415.1 | 16 - 17 | 1985 | 70 | ||
| XM_021253757.1 | XP_021109416.1 | 16 - 17 | 1984 | 70 | ||
More Information
Set Membership: |
|

Arhgef33
Genomic Map