Assay Details
| Gene Symbol: Fabp9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004842003.1 | XP_004842060.1 | 1 - 2 | 112 | 97 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fabp9| Gene Symbol: Fabp9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004842003.1 | XP_004842060.1 | 1 - 2 | 112 | 97 | |