Assay Details
| Gene Symbol: Fbxo31 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004842759.2 | XP_004842816.1 | 8 - 9 | 1321 | 70 | |
| XM_013071462.2 | XP_012926916.2 | 9 - 10 | 1408 | 70 | ||
| XM_013071463.2 | XP_012926917.1 | 9 - 10 | 1113 | 70 | ||
| XM_021256632.1 | XP_021112291.1 | 8 - 9 | 1027 | 70 | ||
More Information
Set Membership: |
|

Fbxo31
Genomic Map