Assay Details
| Gene Symbol: Stx18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004847302.3 | XP_004847359.1 | 5 - 6 | 579 | 71 | |
| XM_013073046.2 | XP_012928500.1 | 4 - 5 | 475 | 71 | ||
| XM_021260300.1 | XP_021115959.1 | 5 - 6 | 901 | 71 | ||
| XM_021260301.1 | XP_021115960.1 | 5 - 6 | 528 | 71 | ||
| XM_021260302.1 | XP_021115961.1 | 5 - 6 | 468 | 71 | ||
| XM_021260303.1 | XP_021115962.1 | 5 - 6 | 449 | 71 | ||
| XM_021260304.1 | XP_021115963.1 | 5 - 6 | 597 | 71 | ||
More Information
Set Membership: |
|

Stx18
Genomic Map