Assay Details
| Gene Symbol: Nek2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004853527.2 | XP_004853584.1 | 4 - 5 | 762 | 80 | |
| XM_004853528.2 | XP_004853585.1 | 4 - 5 | 763 | 80 | ||
| XM_013075336.2 | XP_012930790.1 | 4 - 5 | 762 | 80 | ||
| XM_013075337.2 | XP_012930791.1 | 4 - 5 | 762 | 80 | ||
| XM_021265550.1 | XP_021121209.1 | 4 - 5 | 763 | 80 | ||
| XM_021265551.1 | XP_021121210.1 | 4 - 5 | 639 | 80 | ||
More Information
Set Membership: |
|

Nek2
Genomic Map