Assay Details
| Gene Symbol: Sarm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004856982.2 | XP_004857039.1 | 7 - 8 | 2623 | 62 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sarm1| Gene Symbol: Sarm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004856982.2 | XP_004857039.1 | 7 - 8 | 2623 | 62 | |
Set Membership: |
|