Assay Details
| Gene Symbol: Krt13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004859398.3 | XP_004859455.1 | 6 - 7 | 3519 | 91 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Krt13| Gene Symbol: Krt13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004859398.3 | XP_004859455.1 | 6 - 7 | 3519 | 91 | |