Assay Details
| Gene Symbol: Ecsit | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004865602.2 | XP_004865659.1 | 7 - 8 | 1154 | 74 | |
| XM_021244818.1 | XP_021100477.1 | 7 - 8 | 1148 | 74 | ||
| XM_021244819.1 | XP_021100478.1 | 6 - 7 | 1044 | 74 | ||
| XM_021244820.1 | XP_021100479.1 | 6 - 7 | 1017 | 74 | ||
More Information
Set Membership: |
|

Ecsit
Genomic Map