Assay Details
| Gene Symbol: LOC101707697 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004866847.2 | XP_004866904.1 | 4 - 5 | 876 | 85 | |
| XM_004866848.2 | XP_004866905.1 | 4 - 5 | 777 | 85 | ||
| XM_004866849.2 | XP_004866906.1 | 4 - 5 | 876 | 85 | ||
More Information
Set Membership: |
|

LOC101707697
Genomic Map