Assay Details
| Gene Symbol: Tbpl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004874829.2 | XP_004874886.1 | 4 - 5 | 521 | 73 | |
| XM_004874830.3 | XP_004874887.1 | 4 - 5 | 405 | 73 | ||
More Information
Set Membership: |
|

Tbpl1
Genomic Map