Assay Details
| Gene Symbol: Podxl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004875270.3 | XP_004875327.1 | 7 - 8 | 1536 | 61 | |
| XM_021252500.1 | XP_021108159.1 | 7 - 8 | 1536 | 61 | ||
More Information
Set Membership: |
|

Podxl2
Genomic Map