Assay Details
Gene Symbol: Znf793 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013077442.2 | XP_012932896.1 | 4 - 5 | 688 | 96 | |
XM_021239513.1 | XP_021095172.1 | 3 - 4 | 587 | 96 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_013077442.2, XM_021239513.1 transcripts.
Set Membership: |
![]() ![]() |
