Assay Details
| Gene Symbol: Znf793 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013077442.2 | XP_012932896.1 | 5 - 6 | 729 | 88 | |
| XM_021239513.1 | XP_021095172.1 | 4 - 5 | 628 | 88 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_013077442.2, XM_021239513.1 transcripts.
Set Membership: |
|

Znf793
Genomic Map