Assay Details
Gene Symbol: Cfap65 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021244230.1 | XP_021099889.1 | 17 - 18 | 3201 | 58 | |
XM_021244231.1 | XP_021099890.1 | 17 - 18 | 3200 | 58 | ||
XM_021244232.1 | XP_021099891.1 | 17 - 18 | 3201 | 58 | ||
XM_021244233.1 | XP_021099892.1 | 17 - 18 | 3202 | 58 | ||
XM_021244234.1 | XP_021099893.1 | 13 - 14 | 2673 | 58 | ||
XM_021244235.1 | XP_021099894.1 | 17 - 18 | 3202 | 58 | ||
XM_021244236.1 | XP_021099895.1 | 10 - 11 | 2154 | 58 | ||
XM_021244237.1 | XP_021099896.1 | 10 - 11 | 2144 | 58 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
