Assay Details
| Gene Symbol: LOC101708495 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021255675.1 | XP_021111334.1 | 2 - 3 | 474 | 96 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101708495| Gene Symbol: LOC101708495 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021255675.1 | XP_021111334.1 | 2 - 3 | 474 | 96 | |