Assay Details
| Gene Symbol: LOC101715009 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021265695.1 | XP_021121354.1 | 2 - 3 | 76 | 83 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101715009| Gene Symbol: LOC101715009 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021265695.1 | XP_021121354.1 | 2 - 3 | 76 | 83 | |
Set Membership: |
|