Assay Details
| Gene Symbol: Kmt5a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004843948.2 | XP_004844005.1 | 4 - 5 | 586 | 86 | |
| XM_021257611.1 | XP_021113270.1 | 3 - 4 | 477 | 86 | ||
More Information
Set Membership: |
|

Kmt5a
Genomic Map