Assay Details
| Gene Symbol: Dnajb12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004846243.2 | XP_004846300.1 | 5 - 6 | 817 | 56 | |
| XM_004846244.2 | XP_004846301.1 | 5 - 6 | 817 | 56 | ||
| XM_004846245.2 | XP_004846302.1 | 5 - 6 | 817 | 56 | ||
More Information
Set Membership: |
|

Dnajb12
Genomic Map