Assay Details
| Gene Symbol: LOC101726151 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004849592.1 | XP_004849649.1 | 3 - 4 | 359 | 71 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101726151| Gene Symbol: LOC101726151 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004849592.1 | XP_004849649.1 | 3 - 4 | 359 | 71 | |
Set Membership: |
|