Assay Details
| Gene Symbol: Grxcr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004849786.3 | XP_004849843.1 | 1 - 2 | 386 | 76 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Grxcr1| Gene Symbol: Grxcr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004849786.3 | XP_004849843.1 | 1 - 2 | 386 | 76 | |
Set Membership: |
|