Assay Details
| Gene Symbol: Znf232 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004857327.3 | XP_004857384.1 | 3 - 4 | 932 | 72 | |
| XM_004857328.3 | XP_004857385.1 | 3 - 4 | 757 | 72 | ||
| XM_004857330.3 | XP_004857387.1 | 3 - 4 | 616 | 72 | ||
| XM_004857331.3 | XP_004857388.1 | 2 - 3 | 528 | 72 | ||
| XM_021238246.1 | XP_021093905.1 | 3 - 4 | 771 | 72 | ||
| XM_021238247.1 | XP_021093906.1 | 2 - 3 | 693 | 72 | ||
More Information
Set Membership: |
|

Znf232
Genomic Map