Assay Details
| Gene Symbol: Gdf5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004867871.2 | XP_004867928.1 | 1 - 2 | 1428 | 76 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gdf5| Gene Symbol: Gdf5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004867871.2 | XP_004867928.1 | 1 - 2 | 1428 | 76 | |
Set Membership: |
|