Assay Details
| Gene Symbol: Tymp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013072507.2 | XP_012927961.1 | 4 - 5 | 974 | 61 | |
| XM_013072508.2 | XP_012927962.1 | 5 - 6 | 818 | 61 | ||
| XM_021258710.1 | XP_021114369.1 | 5 - 6 | 732 | 61 | ||
| XM_021258711.1 | XP_021114370.1 | 5 - 6 | 729 | 61 | ||
More Information
Set Membership: |
|

Tymp
Genomic Map