Assay Details
| Gene Symbol: LOC106008570 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021251558.1 | XP_021107217.1 | 2 - 3 | 112 | 70 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC106008570| Gene Symbol: LOC106008570 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021251558.1 | XP_021107217.1 | 2 - 3 | 112 | 70 | |
Set Membership: |
|