Assay Details
| Gene Symbol: LOC106007601 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021256353.1 | XP_021112012.1 | 8 - 9 | 2948 | 99 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC106007601| Gene Symbol: LOC106007601 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021256353.1 | XP_021112012.1 | 8 - 9 | 2948 | 99 | |
Set Membership: |
|