Assay Details
| Gene Symbol: Snrpb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004840668.3 | XP_004840725.1 | 4 - 5 | 604 | 55 | |
| XM_013070686.2 | XP_012926140.1 | 4 - 5 | 604 | 55 | ||
| XM_013070687.2 | XP_012926141.1 | 4 - 5 | 604 | 55 | ||
| XM_021254973.1 | XP_021110632.1 | 4 - 5 | 604 | 55 | ||
More Information
Set Membership: |
|

Snrpb
Genomic Map