Assay Details
| Gene Symbol: Zbtb49 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004847298.2 | XP_004847355.1 | 7 - 8 | 1681 | 87 | |
| XM_021260294.1 | XP_021115953.1 | 7 - 8 | 1759 | 87 | ||
| XM_021260295.1 | XP_021115954.1 | 6 - 7 | 1655 | 87 | ||
| XM_021260296.1 | XP_021115955.1 | 6 - 7 | 1593 | 87 | ||
| XM_021260297.1 | XP_021115956.1 | 5 - 6 | 1486 | 87 | ||
More Information
Set Membership: |
|

Zbtb49
Genomic Map