Assay Details
| Gene Symbol: Slc31a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004849554.3 | XP_004849611.1 | 2 - 3 | 3246 | 64 | |
| XM_004849555.3 | XP_004849612.1 | 2 - 3 | 481 | 64 | ||
| XM_004849556.2 | XP_004849613.1 | 2 - 3 | 241 | 64 | ||
| XM_004849557.2 | XP_004849614.1 | 2 - 3 | 195 | 64 | ||
| XM_013073635.2 | XP_012929089.1 | 2 - 3 | 3324 | 64 | ||
More Information
Set Membership: |
|

Slc31a2
Genomic Map