Assay Details
| Gene Symbol: Slc31a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004849554.3 | XP_004849611.1 | 3 - 4 | 3435 | 84 | |
| XM_004849555.3 | XP_004849612.1 | 3 - 4 | 670 | 84 | ||
| XM_004849556.2 | XP_004849613.1 | 3 - 4 | 430 | 84 | ||
| XM_004849557.2 | XP_004849614.1 | 3 - 4 | 384 | 84 | ||
| XM_013073635.2 | XP_012929089.1 | 3 - 4 | 3513 | 84 | ||
More Information
Set Membership: |
|

Slc31a2
Genomic Map