Assay Details
| Gene Symbol: Pcdh10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004854967.3 | XP_004855024.1 | 3 - 4 | 3677 | 75 | |
| XM_004854969.3 | XP_004855026.1 | 3 - 4 | 3677 | 75 | ||
| XM_021236497.1 | XP_021092156.1 | 3 - 4 | 3677 | 75 | ||
More Information
Set Membership: |
|

Pcdh10
Genomic Map