Assay Details
| Gene Symbol: Pabpc4l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004854970.3 | XP_004855027.1 | 1 | 1418 | 90 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pabpc4l| Gene Symbol: Pabpc4l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004854970.3 | XP_004855027.1 | 1 | 1418 | 90 | |
Set Membership: |
|