Assay Details
| Gene Symbol: Gpr176 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004861530.2 | XP_004861587.1 | 1 - 2 | 958 | 92 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gpr176| Gene Symbol: Gpr176 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004861530.2 | XP_004861587.1 | 1 - 2 | 958 | 92 | |
Set Membership: |
|