Assay Details
| Gene Symbol: Nrl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004864136.3 | XP_004864193.1 | 3 - 4 | 445 | 77 | |
| XM_021243844.1 | XP_021099503.1 | 3 - 4 | 445 | 77 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_004864136.3, XM_021243844.1 transcripts.
Set Membership: |
|

Nrl
Genomic Map