Assay Details
| Gene Symbol: Fsd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004865754.2 | XP_004865811.1 | 11 - 12 | 1544 | 57 | |
| XM_004865756.3 | XP_004865813.1 | 11 - 12 | 1554 | 57 | ||
| XM_004865757.2 | XP_004865814.1 | 11 - 12 | 1439 | 57 | ||
| XM_013065955.2 | XP_012921409.1 | 11 - 12 | 1640 | 57 | ||
| XM_021244985.1 | XP_021100644.1 | 11 - 12 | 1969 | 57 | ||
| XM_021244986.1 | XP_021100645.1 | 11 - 12 | 1470 | 57 | ||
| XM_021244987.1 | XP_021100646.1 | 10 - 11 | 1343 | 57 | ||
More Information
Set Membership: |
|

Fsd1
Genomic Map