Assay Details
| Gene Symbol: Tead2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004866948.2 | XP_004867005.1 | 9 - 10 | 840 | 64 | |
| XM_021246053.1 | XP_021101712.1 | 9 - 10 | 801 | 64 | ||
| XM_021246054.1 | XP_021101713.1 | 8 - 9 | 828 | 64 | ||
| XM_021246056.1 | XP_021101715.1 | 9 - 10 | 841 | 64 | ||
More Information
Set Membership: |
|

Tead2
Genomic Map