Assay Details
| Gene Symbol: Pcsk1n | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004871956.3 | XP_004872013.2 | 2 - 3 | 681 | 77 | |
| XM_004871957.3 | XP_004872014.2 | 2 - 3 | 679 | 77 | ||
More Information
Set Membership: |
|

Pcsk1n
Genomic Map