Assay Details
| Gene Symbol: Slc15a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013069074.2 | XP_012924528.1 | 3 - 4 | 1021 | 57 | |
| XM_021251801.1 | XP_021107460.1 | 3 - 4 | 1023 | 57 | ||
More Information
Set Membership: |
|

Slc15a3
Genomic Map