Assay Details
| Gene Symbol: Runx1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013071314.2 | XP_012926768.1 | 2 - 3 | 228 | 64 | |
| XM_021256212.1 | XP_021111871.1 | 2 - 3 | 228 | 64 | ||
| XM_021256213.1 | XP_021111872.1 | 2 - 3 | 229 | 64 | ||
| XM_021256214.1 | XP_021111873.1 | 2 - 3 | 228 | 64 | ||
More Information
Set Membership: |
|

Runx1
Genomic Map