Assay Details
| Gene Symbol: Slc13a5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013076825.2 | XP_012932279.1 | 1 - 2 | 426 | 82 | |
| XM_021238197.1 | XP_021093856.1 | 2 - 3 | 433 | 82 | ||
| XM_021238198.1 | XP_021093857.1 | 1 - 2 | 427 | 82 | ||
| XM_021238199.1 | XP_021093858.1 | 1 - 2 | 427 | 82 | ||
More Information
Set Membership: |
|

Slc13a5
Genomic Map