Assay Details
| Gene Symbol: Scn1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013077494.2 | XP_012932948.1 | 3 - 4 | 636 | 77 | |
| XM_021239586.1 | XP_021095245.1 | 3 - 4 | 608 | 77 | ||
| XM_021239587.1 | XP_021095246.1 | 3 - 4 | 439 | 77 | ||
| XM_021239588.1 | XP_021095247.1 | 3 - 4 | 433 | 77 | ||
More Information
Set Membership: |
|

Scn1b
Genomic Map