Assay Details
| Gene Symbol: Abi3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021240040.1 | XP_021095699.1 | 7 - 8 | 1137 | 57 | |
| XM_021240041.1 | XP_021095700.1 | 7 - 8 | 1134 | 57 | ||
| XM_021240042.1 | XP_021095701.1 | 7 - 8 | 1086 | 57 | ||
| XM_021240043.1 | XP_021095702.1 | 7 - 8 | 1071 | 57 | ||
| XM_021240044.1 | XP_021095703.1 | 8 - 9 | 961 | 57 | ||
| XM_021240045.1 | XP_021095704.1 | 6 - 7 | 1038 | 57 | ||
More Information
Set Membership: |
|

Abi3
Genomic Map