Assay Details
| Gene Symbol: Pla2g4b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021241805.1 | XP_021097464.1 | 10 - 11 | 1264 | 57 | |
| XM_021241806.1 | XP_021097465.1 | 10 - 11 | 967 | 57 | ||
| XM_021241807.1 | XP_021097466.1 | 10 - 11 | 1274 | 57 | ||
More Information
Set Membership: |
|

Pla2g4b
Genomic Map