Assay Details
| Gene Symbol: Exog | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004835239.3 | XP_004835296.1 | 3 - 4 | 560 | 79 | |
| XM_021259879.1 | XP_021115538.1 | 4 - 5 | 495 | 79 | ||
| XM_021259884.1 | XP_021115543.1 | 4 - 5 | 555 | 79 | ||
| XM_021259889.1 | XP_021115548.1 | 4 - 5 | 499 | 79 | ||
| XM_021259893.1 | XP_021115552.1 | 3 - 4 | 325 | 79 | ||
| XM_021259899.1 | XP_021115558.1 | 2 - 3 | 284 | 79 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_021259899.1 transcripts.
Set Membership: |
|

Exog
Genomic Map