Assay Details
| Gene Symbol: Casp7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004838892.3 | XP_004838949.1 | 2 - 3 | 272 | 64 | |
| XM_004838893.3 | XP_004838950.1 | 2 - 3 | 669 | 64 | ||
| XM_004838894.3 | XP_004838951.1 | 2 - 3 | 222 | 64 | ||
More Information
Set Membership: |
|

Casp7
Genomic Map