Assay Details
| Gene Symbol: Chst4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004842899.2 | XP_004842956.1 | 1 - 2 | 167 | 78 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_004842899.2 transcripts.
Set Membership: |
|

Chst4
Genomic Map