Assay Details
| Gene Symbol: Sh3glb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004849156.3 | XP_004849213.1 | 11 - 12 | 1239 | 73 | |
| XM_004849158.3 | XP_004849215.1 | 10 - 11 | 1176 | 73 | ||
| XM_013073821.2 | XP_012929275.1 | 13 - 14 | 1407 | 73 | ||
| XM_013073822.2 | XP_012929276.1 | 12 - 13 | 1394 | 73 | ||
| XM_013073823.2 | XP_012929277.1 | 12 - 13 | 1391 | 73 | ||
| XM_013073824.2 | XP_012929278.1 | 12 - 13 | 1252 | 73 | ||
| XM_013073826.2 | XP_012929280.1 | 10 - 11 | 1224 | 73 | ||
| XM_013073827.2 | XP_012929281.1 | 9 - 10 | 1161 | 73 | ||
| XM_021261906.1 | XP_021117565.1 | 12 - 13 | 1343 | 73 | ||
| XM_021261907.1 | XP_021117566.1 | 12 - 13 | 1279 | 73 | ||
| XM_021261908.1 | XP_021117567.1 | 14 - 15 | 2240 | 73 | ||
More Information
Set Membership: |
|

Sh3glb2
Genomic Map