Assay Details
| Gene Symbol: Trim44 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004852033.3 | XP_004852090.1 | 1 - 2 | 1155 | 88 | |
| XM_004852034.3 | XP_004852091.1 | 1 - 2 | 1157 | 88 | ||
| XM_021264357.1 | XP_021120016.1 | 2 - 3 | 1309 | 88 | ||
More Information
Set Membership: |
|

Trim44
Genomic Map