Assay Details
| Gene Symbol: Tp53tg5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004858165.2 | XP_004858222.1 | 4 - 5 | 1681 | 68 | |
| XM_004858166.2 | XP_004858223.1 | 3 - 4 | 1616 | 68 | ||
| XM_021239090.1 | XP_021094749.1 | 4 - 5 | 835 | 68 | ||
More Information
Set Membership: |
|

Tp53tg5
Genomic Map